Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Diagnosing Huntington's Using PCR
SDPSCI_ED

Catalog No. S27564 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27564 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27564 Supplier Edvotek™ Supplier No. 1125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Students will conduct a DNA fingerprinting exercise on simulated patient samples to determine if family members are heterozygous or homozygous for Huntington's Disease. Amplified DNA segments are then analyzed by agarose gel electrophoresis.

  • Storage: Some Components Require Freezer Storage
  • Sufficient for 5 complete sets of reactions

Specifications

Grade 11 to Undergraduate
Age 16 to 20
Includes Instructions, PCR EdvoBeads™, LyphoPrimer™ and LyphoTemplate™ DNA, DNA Samples, TE Buffer, UltraSpec-Agarose™, Electrophoresis Buffer (50 x), SYBR™ Safe Stain, FlashBlue™ Liquid Stain, Microcentifuge Tubes, 0.2 mL PCR Tubes, and Wax Beads
Requires Thermal Cycler, Horizontal Gel Electrophoresis Apparatus, Power Supply, Microcentrifuge, UV Transilluminator or UV Photodocumentation System, UV Safety Goggles, White Light Visualization System, Automatic Micropipettes and Tips, Microwave or Hot Plate, 250 mL Flasks/Beakers, Hot Gloves, and Disposable Lab Gloves
Content And Storage Some Components Require Freezer Storage
Class Size 5 Lab Groups
Product Type PCR Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.