Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Diagnosis of White Blood Cell Counts Kit
GREENER_CHOICESCI_ED

Catalog No. s27125 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27125 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27125 Supplier Innovating Science™ Supplier No. IS3106
Only null left
Add to Cart
Add to Cart

Innovating Science™ Diagnosis of White Blood Cell Counts Kit allows students to perform white blood cell counts on five patient samples, and then suggest a possible diagnosis based on the determined cell levels.

  • White blood cells, or leukocytes, only account for approximately 1% of the total number of cells found in blood
  • They are still a critical part of the immune system, protecting the body from foreign invaders such as bacteria and viruses
  • White blood cells counts are often performed as part of a medical diagnosis to determine if the level of white blood cells in a patient’s blood is within, or outside of the normal range
  • In this activity students will perform white blood cell counts on five different patient samples and suggest possible diagnoses based on the white blood cell levels
  • The samples contain safe, non-biological simulated cells that do not require staining
  • Requires microscopes and hemocytometers (not included)
  • Contains enough material for 15 lab groups
  • Includes Teacher’s Guide and Student Study copy masters

Specifications

Certifications/Compliance Non-regulated
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational - Life Science
Includes 5 Simulated Blood Samples (Patient #1, Patient #2, Patient #3, Patient #4, and Patient #5), Teacher Manual, and Student Guide copy masters
Material Cardboard Box
Quantity 1 Kit
Requires Hemocytometers with Coverslips, Microscopes with 10 x Objective (not included)
Weight (English) ∼1 lb.
Weight (Metric) ∼453.6 g
Content And Storage Ambient
Product Line Innovating Science
Dimensions (L x W x H) 9 x 6.5 x 2.75 in.
Class Size 15 Lab Groups
Product Type Diagnosis of White Blood Cell Counts Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.