Learn More
Saf T Pak Inc STP-210xs - Category B Ambient Insulated Shipping System (UN 3373), Extra Small
SDP
Catalog No. NC9509798 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
STP-210xs - Category B Ambient Insulated Shipping System (UN 3373), Extra Small; Maximum Capacity: Four 10mL Vials; 10/Case Reversible CAT B / Exempt Shipper.
Catalog No. NC9509798 Supplier Saf T Pak Inc Supplier No. STP210XS
STP-210xs - Category B Ambient Insulated Shipping System (UN 3373), Extra Small; Maximum Capacity: Four 10mL Vials; 10/Case Reversible CAT B / Exempt Shipper.