Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Signature Diagnostic Test, Cryptosporidium/Giardia

Catalog No. 23200277 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Rapid Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-200-277 Rapid Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-200-277

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sure-Vue™ Signature Diagnostic Test for Cryptosporidium/Giardia provides laboratories the ability to provide a fast accurate result for the two most common intestinal parasite infections utilizing one test.

  • One test that detects and differentiates between Cryptosporidium and Giardia
  • Cryptosporidium CPT Code - 87328, Giardia CPT Code - 87329
  • Proven Performance with high sensitivity (Crypto - 97%/GI - 100%) and specificity (Crypto - 99.5%/GI - 99.1%)
  • Will improve turnaround time for test results in approximately 10 minutes
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C (35.6°F to 46.4°F)
CPT Code Cryptosporidium: 87328, Giardia: 87329
Clia Complexity Moderate
Detectable Analytes Cryptosporidium Oocyst Antigen, Giardia Cyst Antigen
Format Cassette
Sample Type Human Fecal
Sensitivity Crypto 97%, GI 100%
Type Rapid Tests
Includes 30 Test Cards, Package Insert, Sample Treatment Buffer, Reagent A, Reagent B, Specimen Dilution Tubes, Pipettes
Packaging Type Test Kit
Test Time 10 min.
Analysis Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.