Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OraSure Technologies Diagnostics Direct Syphilis Health Check™ Control Set
SDP

Catalog No. 22555215 Shop All OraSure Technologies Products
Change view
Click to view available options
Clia Complexity:
Not Applicable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-555-215 Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-555-215 Supplier OraSure Technologies Supplier No. 10010699
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Syphilis Health CheckTM Control Set is intended for use with the Syphilis Health Check Treponemal Antibody test as a quality control material to monitor the test performance.

  • The Syphilis Health Check Control Set contains syphilis antibodies positive and negative controls diluted in pooled human serum which has been tested and found negative for anti-HIV, anti-HCV and HIV-1 antigen.
  • 1 plastic dropper containing 0.5 mL of Syphilis Positive control.
  • 1 plastic dropper containing 0.5 mL of Syphilis Negative control.
  • Contains 0.09% NaN3 and 0.3% ProClin 300 .

Specifications

Content And Storage 2°C to 8°C (refrigerated)
CE Marker No
Clia Complexity Not Applicable
Detectable Analytes None
Type Kit Controls
Disposable Single-use
For Use With (Equipment) The Syphilis Health Check Treponemal Antibodytest as a quality control material to monitor the test performance.
Includes The External Control Set contains 1 positive and 1 negative syphilis antibody control reagent
Form Liquid Ready-to-Use
Operating Temperature Range 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.