Learn More
MSC Weller Xcelite Diagonal Cutter, 4 in, ESD-Safe, Full-Flush, 1 ea
Catalog No. 501920550
Encompass
This diagonal cutter is an ESD-safe tool designed for full-flush cutting in tight spaces, specifically between closely spaced components. It features a relieved tapered head and cushion grips for comfortable handling, with a cutting capacity of 20 AWG.
  • Designed for full-flush cutting
  • Allows use between closely spaced components
  • Features cushion grips for comfortable handling
  • ESD safe
  • Relieved tapered head
  • Features 20 AWG cutting capacity
Catalog No. 50-192-0550 Supplier MSC Supplier No. 39012687
This diagonal cutter is an ESD-safe tool designed for full-flush cutting in tight spaces, specifically between closely spaced components. It features a relieved tapered head and cushion grips for comfortable handling, with a cutting capacity of 20 AWG.
  • Designed for full-flush cutting
  • Allows use between closely spaced components
  • Features cushion grips for comfortable handling
  • ESD safe
  • Relieved tapered head
  • Features 20 AWG cutting capacity