Learn More
Ampco Diagonal Cutting Pliers with Grips
SDP
Catalog No. 19174205 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
7 in.
Catalog No. Length (English)
19-174-205 7 in.
1 options
Catalog No. 19-174-205 Supplier Ampco Supplier No. P36
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant pliers with grips.
  • Independently tested and found to pose no risk in MRI environments using a static magnetic field of 3-Tesla and a maximum spatial gradient magnetic field of 720-Gauss/cm or less.
  • Conforms to ASTM F2503.
  • Meets the requirements established by ASTM A342.
  • Lenght: 7 in.
Specifications
Product Type Diagonal Cutting Pliers
Length (English) 7 in.
Material Bronze