Learn More
MilliporeSigma™ Supelco™ Diaion™ CR11, Sodium Form
Catalog No. 111015402 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 g
Catalog No. Quantity
11-101-5402 1000 g
1 options
Catalog No. 11-101-5402 Supplier MilliporeSigma™ Supelco™ Supplier No. 13547
Add to Cart
Add to Cart
Weakly acidic cation exchange resin
Diaion™ CR11 is chelating resin with iminodiacetic acid groups. It is considered effective in the removal of chromium trivalent from aqueous solutions. This resin in the Na+ form at 50C is seen to exhibit high sorption capacity.
TRUSTED_SUSTAINABILITY
Specifications
pH 4 to 10
Quantity 1000 g
Product Line Diaion
Capacity (Metric) 0.35 meq/mL by wetted bed volume (calcium)
Diaion is a registered trademark of Mitsubishi Chemical Corp.