Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Diaion™ HP-20

Catalog No. 111016351 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6351 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6351 Supplier MilliporeSigma™ Supelco™ Supplier No. 13606
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Swelling in toluene is 30%

Diaion™ HP-20 is a non-polar copolymer styrene-divynilbenzene adsorbent resin. It is a polymeric adsorbent used in reverse phase chromatography. It has spherical particles with 0.5mm diameter. Diaion™ HP-20 is considered as the most effective adsorbent to eliminate toxic effects in fermentation. It is a synthetic adsorbent used for preconcentration and separation of traces heavy metal ions.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 g
Surface Area (Metric) ˜500 m2/g
Product Line Diaion
Density 1.01 g/mL at 25°C (true wet)(lit.)
CAS 9052-95-3

Diaion is a registered trademark of Mitsubishi Chemical Corp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.