Learn More
MilliporeSigma™ Supelco™ Diaion™ WA30, Free Base
Catalog No. 111004133 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
Catalog No. Quantity
11-100-4133 1 kg
1 options
Catalog No. 11-100-4133 Supplier MilliporeSigma™ Supelco™ Supplier No. 13543
Add to Cart
Add to Cart
Weakly basic anion exchange resin that is best in class for high MW organic acids.
Diaion™ WA30 has a strong decolorization capability. Applications for the resin include: water treatment, pretreatment of corn syrup, beet sugar and dextrose, refining of formalin, glycerine, and enzymes, and as a weak base catalyst. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.
TRUSTED_SUSTAINABILITY
Specifications
pH 0 to 9
Quantity 1 kg
Product Line Diaion
Capacity (Metric) 3.0 meq/g by dry weight
Diaion is a registered trademark of Mitsubishi Chemical Corp.