Learn More
Grainger Mitutoyo Dial Caliper, Stainless Steel, 0 mm to 300 mm, 0.02 mm Graduation, 1 ea
Catalog No. 502767548
Encompass_Preferred
These dial calipers are designed for precision measurements in metalworking, auto repair, and machining applications. Constructed with a durable stainless steel frame, they feature a redesigned dial movement to ensure ultrasmooth sliding and enhanced high-shock protection. The clear, high-readability dial display facilitates accurate readings of outside, inside, depth, and step dimensions.
  • Features a 0 mm to 300 mm measurement range with 0.02 mm graduations
  • Equipped with lock screws for both the dial bezel and the sliding jaw position
  • Includes carbide-tipped jaws for OD measurement
  • Designed with an improved finish on sliding surfaces to enhance operational longevity
  • Supplied in a fitted plastic case and includes a certificate of inspection
Catalog No. 50-276-7548 Supplier Grainger Supplier No. 16X243
These dial calipers are designed for precision measurements in metalworking, auto repair, and machining applications. Constructed with a durable stainless steel frame, they feature a redesigned dial movement to ensure ultrasmooth sliding and enhanced high-shock protection. The clear, high-readability dial display facilitates accurate readings of outside, inside, depth, and step dimensions.
  • Features a 0 mm to 300 mm measurement range with 0.02 mm graduations
  • Equipped with lock screws for both the dial bezel and the sliding jaw position
  • Includes carbide-tipped jaws for OD measurement
  • Designed with an improved finish on sliding surfaces to enhance operational longevity
  • Supplied in a fitted plastic case and includes a certificate of inspection