Learn More
Enterprise Technology Solutions Dial Corporation Dispenser,Touch-free,Refillable,1L Capacity,3/CT,Slate
SDP
Catalog No. 5000831009
Encompass_Preferred
FIT Touch-free Dispenser features a small footprint with subtle curves that complement the aesthetics of any washroom. Enjoy thick, rich foam from a state-of-the-art pump. High-efficiency engine provides an industry-leading precise activations - more than 90,000 - to maximize housekeeping efficiency. Clear side-panel strip provides fill-level visibility without opening. Touch-free unit runs on four D alkaline batteries (included) and can dispense one, two or three shots per activation. Smart-Sensor Technology discontinues activation if hand is removed. Refillable dispenser is made from ABS plastic for maximum durability. Clean Tip Technology cleans the pump tip after each use to minimize drip risk.
Catalog No. 50-008-31009 Supplier Enterprise Technology Solutions Supplier No. DIA16626CT
FIT Touch-free Dispenser features a small footprint with subtle curves that complement the aesthetics of any washroom. Enjoy thick, rich foam from a state-of-the-art pump. High-efficiency engine provides an industry-leading precise activations - more than 90,000 - to maximize housekeeping efficiency. Clear side-panel strip provides fill-level visibility without opening. Touch-free unit runs on four D alkaline batteries (included) and can dispense one, two or three shots per activation. Smart-Sensor Technology discontinues activation if hand is removed. Refillable dispenser is made from ABS plastic for maximum durability. Clean Tip Technology cleans the pump tip after each use to minimize drip risk.