Learn More
Enterprise Technology Solutions Dial Corporation Hand Sanitizer,Refill,Antibacterial,15oz,CL
SDP
Catalog No. 5000790384
Encompass_Preferred
Versa Direct Connect Antibacterial Gel Hand Sanitizer conveniently and effectively kills 99.999 percent of pathogenic bacteria encountered in away-from-home settings in as little as 15 seconds. Dermatologist-tested, hypoallergenic formula is free of fragrances and dyes. Hand sanitizer contains skin conditioners that are gentle on hands even after repeated use. Formula is tested and compatible with latex, vinyl and nitrile gloves. It is NSF E3 listed in the Nonfood Compounds Program. Refill is compatible with Versa Direct Connect Pouch Dispensers (not included).
Catalog No. 50-007-90384 Supplier Enterprise Technology Solutions Supplier No. DIA12258
Versa Direct Connect Antibacterial Gel Hand Sanitizer conveniently and effectively kills 99.999 percent of pathogenic bacteria encountered in away-from-home settings in as little as 15 seconds. Dermatologist-tested, hypoallergenic formula is free of fragrances and dyes. Hand sanitizer contains skin conditioners that are gentle on hands even after repeated use. Formula is tested and compatible with latex, vinyl and nitrile gloves. It is NSF E3 listed in the Nonfood Compounds Program. Refill is compatible with Versa Direct Connect Pouch Dispensers (not included).