Learn More
MSC Chicago Dial Indicator Dial Indicator, Back Plunger, 0.001 in. Graduation, 1 ea.
Catalog No. 5000513608
Encompass
This back plunger set is a dial indicator accessory designed for precision measurement applications within metalworking and industrial environments.
  • Designed for back plunger orientation
  • Features a 1.5500 in. dial diameter
  • Includes 0 to 100 dial reading
  • Provides 0.001000 in. dial graduation
  • Max measurement capacity of 0.2000 in.
  • Constructed with a black finish
  • Manufactured in the USA
Catalog No. 50-005-13608 Supplier MSC Supplier No. 01702851
This back plunger set is a dial indicator accessory designed for precision measurement applications within metalworking and industrial environments.
  • Designed for back plunger orientation
  • Features a 1.5500 in. dial diameter
  • Includes 0 to 100 dial reading
  • Provides 0.001000 in. dial graduation
  • Max measurement capacity of 0.2000 in.
  • Constructed with a black finish
  • Manufactured in the USA