Learn More
Grainger Dial Indicator 0 to 10mm 0-50-0
Catalog No. 502766265
Encompass_Preferred
The Mitutoyo Back Plunger Dial Indicator is a precision measuring instrument used to accurately measure small distances and variations. It has a measuring range of 0-1" (0-25mm) with graduations of 0.001" (0.01mm). The indicator has a carbide-tipped contact point, jeweled movement, and satin chrome finish. It can be used on surfaces not perpendicular to the stem. This high quality dial indicator is ideal for use in inspection, machine shops, tool rooms and more.
Catalog No. 50-276-6265 Supplier Grainger Supplier No. 16X232
The Mitutoyo Back Plunger Dial Indicator is a precision measuring instrument used to accurately measure small distances and variations. It has a measuring range of 0-1" (0-25mm) with graduations of 0.001" (0.01mm). The indicator has a carbide-tipped contact point, jeweled movement, and satin chrome finish. It can be used on surfaces not perpendicular to the stem. This high quality dial indicator is ideal for use in inspection, machine shops, tool rooms and more.