Learn More
Grainger Dial Indicator 0 to 1mm 0-100-0
Catalog No. 502767195
Encompass_Preferred
The Mitutoyo Dial Indicator Flat Back 0-1" is a precision instrument for measuring small distances and variations. It has a 0-1" range with 0.001" graduations and a 1.6" dial face. The flat back design allows it to sit upright on surfaces. It measures in inch and metric units and features a friction thimble to lock the plunger. Made of hardened stainless steel with a satin chrome finish, it comes with a plastic case for protection. This durable dial indicator is ideal for inspection, manufacturing, and machining applications.
Catalog No. 50-276-7195 Supplier Grainger Supplier No. 16X236
The Mitutoyo Dial Indicator Flat Back 0-1" is a precision instrument for measuring small distances and variations. It has a 0-1" range with 0.001" graduations and a 1.6" dial face. The flat back design allows it to sit upright on surfaces. It measures in inch and metric units and features a friction thimble to lock the plunger. Made of hardened stainless steel with a satin chrome finish, it comes with a plastic case for protection. This durable dial indicator is ideal for inspection, manufacturing, and machining applications.