Learn More
Grainger Dial Indicator 0 to 1mm 50-0-50
Catalog No. 502766353
Encompass_Preferred
The Mitutoyo Horizontal Dial Test Indicator is a precision measuring instrument used to accurately measure small distances and variations. It has a measuring range of 0-0.5" (0-12.7mm) with graduations of 0.001" (0.01mm). The indicator has a horizontal dial face with a bezel clamp to mount on surfaces. It can measure variations in travel, vibration, runout, deflection and more. The indicator has a carbide contact point and comes with a NIST traceable certificate. It is made in Japan by Mitutoyo, a leading manufacturer of precision measuring tools.
Catalog No. 50-276-6353 Supplier Grainger Supplier No. 16X225
The Mitutoyo Horizontal Dial Test Indicator is a precision measuring instrument used to accurately measure small distances and variations. It has a measuring range of 0-0.5" (0-12.7mm) with graduations of 0.001" (0.01mm). The indicator has a horizontal dial face with a bezel clamp to mount on surfaces. It can measure variations in travel, vibration, runout, deflection and more. The indicator has a carbide contact point and comes with a NIST traceable certificate. It is made in Japan by Mitutoyo, a leading manufacturer of precision measuring tools.