Learn More
Grainger Dial Inflator Gauge 3 ft
Catalog No. 502751427
Encompass_Preferred
The Haltec Dial Inflator Gauge is a dial-type tire pressure gauge used to measure tire pressure. It has a 2.5 inch dial with a dual calibrated scale that reads 0-60 psi and 0-4.1 bar. The gauge has a rubber hose and chuck that connects to the tire valve stem. To use, press the chuck onto the valve stem and read the pressure on the dial. The gauge has an all steel construction in a chrome plated finish. It can help maintain proper tire inflation for safety, maximum tire life and fuel economy. The gauge measures 5 inches long.
Catalog No. 50-275-1427 Supplier Grainger Supplier No. 140Z43
The Haltec Dial Inflator Gauge is a dial-type tire pressure gauge used to measure tire pressure. It has a 2.5 inch dial with a dual calibrated scale that reads 0-60 psi and 0-4.1 bar. The gauge has a rubber hose and chuck that connects to the tire valve stem. To use, press the chuck onto the valve stem and read the pressure on the dial. The gauge has an all steel construction in a chrome plated finish. It can help maintain proper tire inflation for safety, maximum tire life and fuel economy. The gauge measures 5 inches long.