Learn More
Grainger Dial Scale SS 50 lb Weight Cap. Silver
Catalog No. 502750935
Encompass_Preferred
The Dial Scale SS-50 is a 50 lb capacity stainless steel mechanical dial scale with a 6 inch diameter dial face. It has a zero adjust knob and features a sturdy steel base with rubber feet. The scale weighs 5 lbs and measures 11 x 7 x 2 inches. It can be used for weighing items in industrial, commercial, or home settings. The scale is made in the USA by Dial Corporation, a leader in precision measurement instruments. It comes with a 5 year limited warranty.
Catalog No. 50-275-0935 Supplier Grainger Supplier No. 14F322
The Dial Scale SS-50 is a 50 lb capacity stainless steel mechanical dial scale with a 6 inch diameter dial face. It has a zero adjust knob and features a sturdy steel base with rubber feet. The scale weighs 5 lbs and measures 11 x 7 x 2 inches. It can be used for weighing items in industrial, commercial, or home settings. The scale is made in the USA by Dial Corporation, a leader in precision measurement instruments. It comes with a 5 year limited warranty.