Learn More
MSC Rubbermaid Commercial Refrigerator/Freezer Thermometer, -20-80 °F, Dial, NSF Certified
Catalog No. 502836364
Encompass
The Refrigerator/Freezer Thermometer is designed to keep food safe by providing accurate temperature monitoring. It features an easy-to-read dial and offers flexible installation options, allowing it to hang or stand. This thermometer reads both Fahrenheit and Celsius, with a temperature range of -20 to 80 degrees Fahrenheit. It is also NSF and REACH certified, ensuring compliance with food safety and chemical regulations.
  • Keeps food safe
  • Features easy-to-read dial
  • Hangs or stands for easy installation
  • Reads both Fahrenheit and Celsius
  • Ranges from -20 to 80 degrees Fahrenheit
  • NSF certified
  • REACH compliant
Catalog No. 50-283-6364 Supplier MSC Supplier No. 57801318
The Refrigerator/Freezer Thermometer is designed to keep food safe by providing accurate temperature monitoring. It features an easy-to-read dial and offers flexible installation options, allowing it to hang or stand. This thermometer reads both Fahrenheit and Celsius, with a temperature range of -20 to 80 degrees Fahrenheit. It is also NSF and REACH certified, ensuring compliance with food safety and chemical regulations.
  • Keeps food safe
  • Features easy-to-read dial
  • Hangs or stands for easy installation
  • Reads both Fahrenheit and Celsius
  • Ranges from -20 to 80 degrees Fahrenheit
  • NSF certified
  • REACH compliant