Learn More
Electron Microscopy Sciences Dial Vernier Caliper Metric Scale
SDP
Catalog No. 501899075
Encompass_Preferred
These precision vernier calipers are molded in corrosion resistant fiberglass-filled super-polyamide nylon alloy. They are excellent tools due to there easy reading scale (black numbers on white background) and dials (light weight, fine adjustment thumb roller). Its parallel jaws are 1.57" (40 mm) deep, and its reading scale does not create a parallax. Its inside-outside jaws and round metal rod depth gauge add to its value.
B Dial type, 0.1 mm dial, 1.3" (33 mm) diameter, maximum reading 150 mm, 10 mm bar scale.
Catalog No. 50-189-9075 Supplier Electron Microscopy Sciences Supplier No. 62065B
These precision vernier calipers are molded in corrosion resistant fiberglass-filled super-polyamide nylon alloy. They are excellent tools due to there easy reading scale (black numbers on white background) and dials (light weight, fine adjustment thumb roller). Its parallel jaws are 1.57" (40 mm) deep, and its reading scale does not create a parallax. Its inside-outside jaws and round metal rod depth gauge add to its value.
B Dial type, 0.1 mm dial, 1.3" (33 mm) diameter, maximum reading 150 mm, 10 mm bar scale.