Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diallyl carbonate

Catalog No. 2812395G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-123-95G 5 g
28-123-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-123-95G Supplier Sigma Aldrich Supplier No. 2812395G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diallyl carbonate is an acrylating agent. Allyl carbonates are widely employed in Tsuji-Trost allylation, generating carbanion, boronate, phosphide, amide and alkoxide, nucleophiles in situ and improving reaction rates over allyl acetate. Allyl carbonates are also of considerable interest for the intramolecular decarboxylative assymetric couplings.Transesterification of diallyl carbonate with glycerol in the presence of a catalyst (Pd/PPh3), generates glycerol carbonate.

Specifications

CAS 15022-08-9
Density 0.991 g/mL (at 25°C)
Boiling Point 95°C to 97°C (60 mmHg)
Molecular Formula C7H10O3
Refractive Index n20/D 1.428 (literature)
Linear Formula (H2C=CHCH2O)2 CO
Quantity 5 g
MDL Number MFCD00008645
Molecular Weight (g/mol) 142.15
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.