Learn More
Medchemexpress LLC Diallyl disulfide | 2179-57-9 | MFCD00008656 | 98.4% | 146.27 g/mol | C6H10S2 | 10 MG
SDP
Catalog No. 5000301590
Encompass_Preferred
Diallyl disulfide is an organosulfur compound derived from garlic oil. It is reported as an orally active human squalene monooxygenase inhibitor (IC50 ≈400 μM) and has been investigated for anti-inflammatory, antioxidant, antidepressant, and antitumor activities.
  • Organosulfur compound with molecular formula C6H10S2 and molecular weight 146.27 g/mol.
  • Orally active squalene monooxygenase inhibitor (IC50 ≈400 μM).
  • Reported biological activities include anti-inflammatory, antioxidant, antidepressant, and antitumor effects.
  • High purity analytical grade available (≈98.4% per data sheet).
  • Storage: store at 4°C under nitrogen; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-003-01590 Supplier Medchemexpress LLC Supplier No. HYW01563510MG
Diallyl disulfide is an organosulfur compound derived from garlic oil. It is reported as an orally active human squalene monooxygenase inhibitor (IC50 ≈400 μM) and has been investigated for anti-inflammatory, antioxidant, antidepressant, and antitumor activities.
  • Organosulfur compound with molecular formula C6H10S2 and molecular weight 146.27 g/mol.
  • Orally active squalene monooxygenase inhibitor (IC50 ≈400 μM).
  • Reported biological activities include anti-inflammatory, antioxidant, antidepressant, and antitumor effects.
  • High purity analytical grade available (≈98.4% per data sheet).
  • Storage: store at 4°C under nitrogen; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).