Learn More
Cayman Chemical Diallyl Disulfide, Purity: ≥80%, CAS: 2179-57-9, 500 mg
SDP
Catalog No. 502889155
Encompass_Preferred
An organosulfur compound derived from allicin; converted, in the presence of thiols, to the gaseous mediator H2S; serves as a thiol-dependent H2S donor in biological systems
Catalog No. 50-288-9155 Supplier Cayman Chemical Supplier No. 10012582500 MG
An organosulfur compound derived from allicin; converted, in the presence of thiols, to the gaseous mediator H2S; serves as a thiol-dependent H2S donor in biological systems