Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl disulfide, ≥96.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018922 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8922 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8922 Supplier MilliporeSigma Supelco Supplier No. 306481ML
Add to Cart
Add to Cart

Diallyl disulfide is an oil-soluble organosulfer compound found in garlic, which is known to exert chemopreventive activity against colon, lung, and skin cancer.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 2179-57-9
Density 1.0 g/mL (at 20°C)
Flash Point 62°C - Closed Cup
Linear Formula CH2=CHCH2 SSCH2 CH=CH2
MDL Number MFCD00008656
Synonym Allyl disulfide
Type Analytical Standard
Quantity 1 mL
Formula Weight 146.27
Percent Purity ≥96.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.