Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl Isocyanurate 98.0+%, TCI America™
SDP

Catalog No. I030425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I030425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I030425G Supplier TCI America Supplier No. I030425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6294-79-7
Molecular Formula C9H11N3O3
Molecular Weight (g/mol) 209.205
MDL Number MFCD00059763
InChI Key UCBVELLBUAKUNE-UHFFFAOYSA-N
Synonym Isocyanuric Acid Diallyl Ester
PubChem CID 94974
IUPAC Name 1,3-bis(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
SMILES C=CCN1C(=O)NC(=O)N(C1=O)CC=C

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 209.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diallyl Isocyanurate
EINECSNumber (5)-5912
RTECSNumber XZ1835000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.