Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl Maleate, TCI America™
SDP

Catalog No. M0008500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0008500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0008500G Supplier TCI America Supplier No. M0008500G

missing translation for 'validMainframeMsg'

Specifications

CAS 999-21-3
Melting Point -47°C
Color Yellow
Boiling Point 154°C
Molecular Formula C10H12O4
MDL Number MFCD00010763
UN Number 2810
Quantity 500 g
Synonym diallyl fumarate, fumaric acid, diallyl ester, unii-k852lk4c5p, 2-butenedioic acid e-, di-2-propenyl ester, 2-butenedioic acid 2e-, di-2-propenyl ester, butenedioic acid diallyl ester, 1,4-bis prop-2-en-1-yl 2e-but-2-enedioate, 2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester, diallyl malleate, 1,4-bis prop-2-en-1-yl but-2-enedioate
InChI Key ZPOLOEWJWXZUSP-WAYWQWQTSA-N
SMILES C=CCOC(=O)C=CC(=O)OCC=C
IUPAC Name bis(prop-2-enyl) (Z)-but-2-enedioate
Molecular Weight (g/mol) 196.202
PubChem CID 5354266
Formula Weight 196.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl Maleate
Show More Show Less
EINECSNumber (2)-1110
RTECSNumber ON0700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.