Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl phthalate, MilliporeSigma™ Supelco™

Catalog No. 111020439 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0439 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0439 Supplier MilliporeSigma Supelco Supplier No. 36925250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 725°F
CAS 131-17-9
Vapor Pressure 2.3 mmHg (150°C)
Vapor Density 8.3 (vs. air)
Density 1.121 g/mL (at 25°C)
Flash Point 166°C - Closed Cup
Physical Form Neat
Linear Formula C6H4-1,2-(CO2 CH2 CH=CH2)2
MDL Number MFCD00008646
Boiling Point 165°C to 167°C (5 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 246.26
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.