Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl Propyl Isocyanurate (stabilized with BHT) 97.0+%, TCI America™
SDP

Catalog No. I04095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04095G Supplier TCI America Supplier No. I04095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5320-25-2
Molecular Formula C12H17N3O3
Molecular Weight (g/mol) 251.286
MDL Number MFCD00144514
InChI Key ZIJDEADTCQKATN-UHFFFAOYSA-N
Synonym Isocyanuric Acid Diallyl Propyl Ester
PubChem CID 12612564
IUPAC Name 1,3-bis(prop-2-enyl)-5-propyl-1,3,5-triazinane-2,4,6-trione
SMILES CCCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 251.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl Propyl Isocyanurate (stabilized with BHT)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.