Learn More
Cayman Chemical Diallyl Trisulfide, Purity: ≥95%, CAS: 2050-87-5, 100 mg
SDP
Catalog No. 502889157
Encompass_Preferred
An organic polysulfide compound found in garlic that acts as an H2S donor; reduces the survival of prostate cancer PC-3 cells (IC50 = 22 μM) and inhibits the growth of human colon adenocarcinoma HCT15 cells (IC50 = 11.5 μM); suppresses the growth of PC-3 xenografts in vivo in mice and induces vascular smooth muscle relaxation.
Catalog No. 50-288-9157 Supplier Cayman Chemical Supplier No. 10012577100 MG
An organic polysulfide compound found in garlic that acts as an H2S donor; reduces the survival of prostate cancer PC-3 cells (IC50 = 22 μM) and inhibits the growth of human colon adenocarcinoma HCT15 cells (IC50 = 11.5 μM); suppresses the growth of PC-3 xenografts in vivo in mice and induces vascular smooth muscle relaxation.