Learn More
Biodesign Inc of Ny Dialysis Tubing, 14000 molecular weight cut-off, 15.5 mm wet diameter, 100 foot roll
Catalog No. NC2545580
Encompass
Dialysis Tubing, 14000 molecular weight cut-off, 15.5 mm wet diameter, 100 foot roll
Catalog No. NC2545580 Supplier Biodesign Inc of Ny Supplier No. D002
Dialysis Tubing, 14000 molecular weight cut-off, 15.5 mm wet diameter, 100 foot roll