Learn More
Biodesign Inc of Ny Dialysis Tubing, 14000 molecular weight cut-off, 21 mm wet diameter, 100 foot roll
Catalog No. NC2545588
Encompass
Dialysis Tubing, 14000 molecular weight cut-off, 21 mm wet diameter, 100 foot roll
Catalog No. NC2545588 Supplier Biodesign Inc of Ny Supplier No. D004
Dialysis Tubing, 14000 molecular weight cut-off, 21 mm wet diameter, 100 foot roll