Learn More
Biodesign Inc of Ny Dialysis Tubing, 14000 molecular weight cut-off, 28 mm wet diameter, 100 foot roll
Catalog No. NC2545596
Encompass
Dialysis Tubing, 14000 molecular weight cut-off, 28 mm wet diameter, 100 foot roll
Catalog No. NC2545596 Supplier Biodesign Inc of Ny Supplier No. D006
Dialysis Tubing, 14000 molecular weight cut-off, 28 mm wet diameter, 100 foot roll