Learn More
Biodesign Inc of Ny Dialysis Tubing, 8000 molecular weight cut-off, 6.4 mm wet diameter, 100 foot roll
Catalog No. NC2545617
Encompass
Dialysis Tubing, 8000 molecular weight cut-off, 6.4 mm wet diameter, 100 foot roll
Catalog No. NC2545617 Supplier Biodesign Inc of Ny Supplier No. D100
Dialysis Tubing, 8000 molecular weight cut-off, 6.4 mm wet diameter, 100 foot roll