Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Dialysis Tubing
SCI_ED

Catalog No. S02420 Shop All Aldon Products
Change view
Click to view available options
Diameter (Metric):
10 mm
Length (English):
100 ft.
Width (Metric):
6.4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (English) Width (Metric)
S02420 10 mm 100 ft. 6.4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02420 Supplier Innovating Science™ Supplier No. IS35044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dialysis tubing of various sizes.

The semi-permeable membrane allows for selective transport within a solution.

Specifications

Product Type Dialysis Tubing
MWCO 7 kDa
Format Tubing
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (Metric) 6.4 mm
Sample Volume (Metric) 2.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.