Learn More
Sigma Aldrich Fine Chemicals Biosciences Dialysis tubing cellulose
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D9527100FT
Encompass
Typical molecular weight cut-off 14 000.
Catalog No. 50-290-0314