Learn More
Sigma Aldrich Fine Chemicals Biosciences Dialysis tubing cellulose membrane, 100 ft
Catalog No. NC0708716
Encompass
Dialysis tubing cellulose membrane, 100 ft. Typical molecular weight cut-off = 14,000.
Catalog No. NC0708716 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D9777100FT
Dialysis tubing cellulose membrane, 100 ft. Typical molecular weight cut-off = 14,000.