Learn More
Sigma Aldrich Fine Chemicals Biosciences Dialysis tubing cellulose
Catalog No. 502901111
Encompass
Typical molecular weight cut-off 14 000.
Catalog No. 50-290-1111 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D9402100FT
Typical molecular weight cut-off 14 000.