Learn More
Biodesign Inc of Ny Dialysis Tubing closure clamps, for dialysis tubing up to 50mm wet diameter, plastic, doubled bar sealing for highly assured closure, re-usable hundred of times, 10 per pack
Catalog No. NC1442481
Encompass
Dialysis Tubing closure clamps, for dialysis tubing up to 50mm wet diameter, plastic, doubled bars for highly assured closure, re-usable hundred of times, 10 per pack
Catalog No. NC1442481 Supplier Biodesign Inc of Ny Supplier No. D805
Dialysis Tubing closure clamps, for dialysis tubing up to 50mm wet diameter, plastic, doubled bars for highly assured closure, re-usable hundred of times, 10 per pack