Learn More
Biodesign Inc of Ny Dialysis Tubing closure clamps, for dialysis tubing up to 21 mm wet diameter, plastic, doubled bar sealing for highly assured closure, re-usable for hundred of times, 10 per pack
Catalog No. NC2545656
Encompass
Dialysis Tubing closure clamps, for dialysis tubing up to 21mm wet diameter, plastic, doubled bars for highly assured closure, re-usable hundred of times, 10 per pack
Catalog No. NC2545656 Supplier Biodesign Inc of Ny Supplier No. D801
Dialysis Tubing closure clamps, for dialysis tubing up to 21mm wet diameter, plastic, doubled bars for highly assured closure, re-usable hundred of times, 10 per pack