Learn More
Biodesign Inc of Ny Dialysis Tubing closure clamps, for dialysis tubing up to 29mm wet diameter, plastic, doubled bar sealing for highly assured closure, re-usable hundred of times, 10 per pack
Catalog No. NC2545662
Encompass
Dialysis Tubing closure clamps, for dialysis tubing up to 29mm wet diameter, plastic, doubled bars for highly assured closure, re-usable hundred of times, 10 per pack
Catalog No. NC2545662 Supplier Biodesign Inc of Ny Supplier No. D803
Dialysis Tubing closure clamps, for dialysis tubing up to 29mm wet diameter, plastic, doubled bars for highly assured closure, re-usable hundred of times, 10 per pack