Learn More
Sigma Aldrich Fine Chemicals Biosciences Dialysis tubing high reten
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D0655100FT
Encompass
Retains 99% of cytochrome c (M.W. 12 400) in SOLUTIONon over a 10-hour period.
Catalog No. 50-290-1354