Learn More
Sigma Aldrich Fine Chemicals Biosciences Dialysis tubing high reten
Catalog No. 502900552
Encompass
Retains 99% of cytochrome c (M.W. 12 400) in SOLUTIONon over a 10-hour period.
Catalog No. 50-290-0552 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D0530100FT
Retains 99% of cytochrome c (M.W. 12 400) in SOLUTIONon over a 10-hour period.