Learn More
Electron Microscopy Sciences DiATOME Cryo Diamond Knife Resharpen Dry 2.1mm 35° Knife angle
SDP
Catalog No. 501898747
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 2.1 mm
Knife angle: 35°
Catalog No. 50-189-8747 Supplier Electron Microscopy Sciences Supplier No. 21CDLR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 2.1 mm
Knife angle: 35°