Learn More
Electron Microscopy Sciences DiATOME Cryo Diamond Knife Resharpen Dry 2.1mm 45° Knife angle
SDP
Catalog No. 501898750
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 2.1 mm
Knife angle: 45°
Catalog No. 50-189-8750 Supplier Electron Microscopy Sciences Supplier No. 21CDSR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 2.1 mm
Knife angle: 45°