Learn More
Electron Microscopy Sciences DiATOME Cryo Immuno Diamond Knife 3.0 mm, 35° Knife angle
SDP
Supplier:  Electron Microscopy Sciences DCIMM3530
Encompass
The first cryo knife with a diamond platform, guarantees the best possible sectioning for cryo-immuno-chemistry. The diamond platform guarantees easy and gentle section collection. Sections are collected directly from the diamond surface using a loop and a sucrose/methylcellulose droplet (Refs. Liou, Peters). The 35 degree cutting angle leads to a considerable reduction in mechanical stresses and therefore to improved structure preservation in sucrose-protected samples.
Size: 3.0 mm
Knife angle: 35°
Catalog No. 50-190-0156