Learn More
Electron Microscopy Sciences DiATOME Cryo Diamond Knife Resharpen Wet 1.8 mm 45° Knife angle
SDP
Catalog No. 501902330
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 1.8 mm
Knife angle: 45°
Catalog No. 50-190-2330 Supplier Electron Microscopy Sciences Supplier No. 18CWSR
Add to Cart
Add to Cart
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 1.8 mm
Knife angle: 45°