Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Exchange Dry 2.7mm 35° Knife angle
SDP
Catalog No. 501902421
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm. Exchange any diamond knife for a new DiATOME Ultra Diamond Knife. It does not matter what size or boat style that is traded in.
Size: 2.7 mm
Knife angle: 35°
Catalog No. 50-190-2421 Supplier Electron Microscopy Sciences Supplier No. 27ULDE
The optimal thickness range for cutting with these knives is between 30-150 nm. Exchange any diamond knife for a new DiATOME Ultra Diamond Knife. It does not matter what size or boat style that is traded in.
Size: 2.7 mm
Knife angle: 35°