Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Resharpen 2.4 mm, 35° Knife angle
SDP
Catalog No. 501898771
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 2.4 mm
Knife angle: 35°
Catalog No. 50-189-8771 Supplier Electron Microscopy Sciences Supplier No. 24ULR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 2.4 mm
Knife angle: 35°