Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Resharpen 4.0 mm, 35° Knife angle
SDP
Catalog No. 501898850
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 4.0 mm
Knife angle: 35°
Catalog No. 50-189-8850 Supplier Electron Microscopy Sciences Supplier No. 40ULR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 4.0 mm
Knife angle: 35°